siRNA Details
| Gene Symbol: C6orf47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021184.3 | NP_067007.3 | 1 | 1074 | ||
| GenBank mRNA | AJ245433.1 | - | 1 | 1035 | ||
| AK315349.1 | - | 1 | 763 | |||
| BC012950.1 | - | 1 | 1074 | 3875402 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C6orf47| Gene Symbol: C6orf47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021184.3 | NP_067007.3 | 1 | 1074 | ||
| GenBank mRNA | AJ245433.1 | - | 1 | 1035 | ||
| AK315349.1 | - | 1 | 763 | |||
| BC012950.1 | - | 1 | 1074 | 3875402 | ||