siRNA Details
| Gene Symbol: KRTAP5-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021046.2 | NP_066384.2 | 1 | 1152 | ||
| GenBank mRNA | BX099510.1 | - | 1 | 487 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP5-8| Gene Symbol: KRTAP5-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021046.2 | NP_066384.2 | 1 | 1152 | ||
| GenBank mRNA | BX099510.1 | - | 1 | 487 | ||