siRNA Details
| Gene Symbol: TP53INP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005260476.2 | XP_005260533.1 | 5 | 1440 | ||
| XM_006723841.2 | XP_006723904.1 | 3 | 1086 | |||
| XM_011528952.2 | XP_011527254.1 | 5 | 1501 | |||
| XM_011528953.2 | XP_011527255.1 | 4 | 1381 | |||
| GenBank mRNA | AJ297792.1 | - | 5 | 1448 | ||
| AL137597.1 | - | 1 | 268 | |||
| AY422170.1 | - | 5 | 1457 | |||
| BC035639.1 | - | 5 | 1346 | 5547220 | ||

TP53INP2
Genomic Map