siRNA Details
| Gene Symbol: SRPRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021203.3 | NP_067026.3 | 7 | 859 | ||
| GenBank mRNA | AF086914.1 | - | 6 | 597 | ||
| AF141882.3 | - | 6 | 590 | |||
| AK027307.1 | - | 3 | 1102 | |||
| AK027525.1 | - | 6 | 590 | |||
| AK075531.1 | - | 6 | 628 | |||
| BC063001.1 | - | 6 | 602 | 5185997 | ||
| BC065299.1 | - | 6 | 574 | 6146939 | ||

SRPRB
Genomic Map