siRNA Details
| Gene Symbol: ENOPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001292017.1 | NP_001278946.1 | 6 | 1030 | ||
| NM_021204.4 | NP_067027.1 | 6 | 1059 | |||
| NR_120457.1 | 5 | 827 | ||||
| GenBank mRNA | AF113125.1 | - | 6 | 930 | ||
| AK022656.1 | - | 6 | 925 | |||
| AL110167.1 | - | 2 | 160 | |||
| BC001317.1 | - | 5 | 645 | |||
| BC065815.1 | - | 6 | 897 | 6526329 | ||
| CR457141.1 | - | 6 | 709 | |||

ENOPH1
Genomic Map