siRNA Details
| Gene Symbol: RPRD1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021215.3 | NP_067038.1 | 6 | 1188 | ||
| XM_005260477.3 | XP_005260534.1 | 6 | 1272 | |||
| XM_005260480.4 | XP_005260537.1 | 6 | 1409 | |||
| XM_011528954.2 | XP_011527256.1 | 6 | 1272 | |||
| XM_017027990.1 | XP_016883479.1 | 6 | 1272 | |||
| XM_017027991.1 | XP_016883480.1 | 5 | 1138 | |||
| GenBank mRNA | AK092567.1 | - | 1 | 143 | ||
| AK312468.1 | - | 6 | 948 | |||
| BC033629.1 | - | 6 | 972 | 5223673 | ||
| DQ372938.1 | - | 6 | 1061 | |||

RPRD1B
Genomic Map