siRNA Details
| Gene Symbol: ZNF77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021217.2 | NP_067040.1 | 4 | 738 | ||
| XM_017027081.1 | XP_016882570.1 | 3 | 601 | |||
| XM_017027082.1 | XP_016882571.1 | 4 | 763 | |||
| GenBank mRNA | AK290979.1 | - | 4 | 674 | ||
| BC043354.1 | - | 4 | 699 | 6014754 | ||
| BC062561.1 | - | 4 | 700 | 6644579 | ||

ZNF77
Genomic Map