siRNA Details
| Gene Symbol: OVOL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303461.1 | NP_001290390.1 | 3, 4 | 717 | ||
| NM_001303462.1 | NP_001290391.1 | 2, 3 | 306 | |||
| NM_021220.3 | NP_067043.2 | 3, 4 | 766 | |||
| GenBank mRNA | AK022284.1 | - | 3, 4 | 761 | ||
| AL079276.1 | - | 2, 3 | 346 | |||
| BC006148.1 | - | 3, 4 | 766 | 3959436 | ||
| BE791524.1 | - | 3, 4 | 579 | |||
| BM563561.1 | - | 2, 3 | 306 | |||
| BT007295.1 | - | 3, 4 | 518 | |||

OVOL2
Genomic Map