siRNA Details
| Gene Symbol: MYL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021223.2 | NP_067046.1 | 1 | 100 | ||
| XM_005249817.4 | XP_005249874.1 | 2 | 174 | |||
| XM_011515463.2 | XP_011513765.1 | 2 | 175 | |||
| XM_011515464.2 | XP_011513766.1 | 2 | 174 | |||
| XM_011515465.2 | XP_011513767.1 | 2 | 174 | |||
| XM_017012478.1 | XP_016867967.1 | 2 | 173 | |||
| XM_017012479.1 | XP_016867968.1 | 2 | 141 | |||
| GenBank mRNA | AK311869.1 | - | 2 | 115 | ||
| BC027915.1 | - | 1 | 96 | 5223298 | ||
| M94547.1 | - | 1 | 100 | |||
| W17098.1 | - | 2 | 101 | |||

MYL7
Genomic Map