siRNA Details
| Gene Symbol: OPRPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302807.1 | NP_001289736.1 | 4 | 760 | ||
| NM_021225.4 | NP_067048.4 | 3 | 704 | |||
| NR_126503.1 | 2 | 638 | ||||
| GenBank mRNA | CD721906.1 | - | 1 | 191 | ||
| CD723628.1 | - | 1 | 203 | |||
| CK429697.1 | - | 2 | 555 | |||
| CK430664.1 | - | 1 | 139 | |||
| S83198.1 | - | 3 | 611 | |||

OPRPN
Genomic Map