siRNA Details
| Gene Symbol: OPRPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302807.1 | NP_001289736.1 | 4 | 678 | ||
| NM_021225.4 | NP_067048.4 | 3 | 622 | |||
| NR_126503.1 | 2 | 556 | ||||
| GenBank mRNA | CD721906.1 | - | 1 | 109 | ||
| CD723628.1 | - | 1 | 121 | |||
| CK429697.1 | - | 2 | 473 | |||
| CK430664.1 | - | 1 | 57 | |||
| S83198.1 | - | 3 | 528 | |||

OPRPN
Genomic Map