siRNA Details
| Gene Symbol: DNASE2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021233.2 | NP_067056.2 | 6 | 916 | ||
| NM_058248.1 | NP_490649.1 | 4 | 666 | |||
| XM_011541878.2 | XP_011540180.1 | 4 | 818 | |||
| GenBank mRNA | AF274571.1 | - | 6 | 916 | ||
| AF333389.1 | - | 4 | 666 | |||
| BC065182.1 | - | 4 | 560 | |||
| BC080621.1 | - | 4 | 554 | |||
| BC126136.1 | - | 6 | 907 | 8991852 | ||
| BC126138.1 | - | 6 | 907 | 8991854 | ||

DNASE2B
Genomic Map