siRNA Details
| Gene Symbol: SELK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021237.3 | NP_067060.2 | 3 | 356 | ||
| GenBank mRNA | AF085359.1 | - | 3 | 312 | ||
| AF161415.1 | - | 3 | 345 | |||
| AF537132.1 | - | 3 | 248 | |||
| AK289594.1 | - | 3 | 249 | |||
| BC013162.1 | - | 3 | 252 | 4345594 | ||
| BM762385.1 | - | 3 | 382 | |||

SELK
Genomic Map