siRNA Details
| Gene Symbol: RRAGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021244.4 | NP_067067.1 | 4 | 950 | ||
| XM_005248755.4 | XP_005248812.1 | 4 | 1144 | |||
| XM_011536022.2 | XP_011534324.1 | 4 | 722 | |||
| GenBank mRNA | AF272036.1 | - | 4 | 733 | ||
| AK289799.1 | - | 4 | 875 | |||
| AL137502.1 | - | 3 | 448 | |||
| BC003088.1 | - | 4 | 612 | 3536088 | ||

RRAGD
Genomic Map