siRNA Details
Gene Symbol: HRH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143828.1 | NP_001137300.1 | 2 | 797 | ||
NM_001160166.1 | NP_001153638.1 | 2 | 897 | |||
NM_021624.3 | NP_067637.2 | 3 | 1061 | |||
GenBank mRNA | AB044934.1 | - | 3 | 1015 | ||
AB045370.1 | - | 3 | 1024 | |||
AF307973.1 | - | 3 | 961 | |||
AF312230.1 | - | 3 | 1061 | |||
AF325356.1 | - | 3 | 961 | |||
AF329449.1 | - | 3 | 961 | |||
AJ298292.1 | - | 3 | 961 | |||
AY008280.1 | - | 3 | 961 | |||
AY136745.1 | - | 3 | 961 | |||
BC069136.1 | - | 3 | 1061 | 7216992 | ||
BC112348.1 | - | 3 | 1021 | 40006540 | ||
DQ835186.1 | - | 2 | 697 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
