siRNA Details
| Gene Symbol: LGR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017403.1 | NP_001017403.1 | 10 | 965 | ||
| NM_001017404.1 | NP_001017404.1 | 8 | 548 | |||
| NM_021636.2 | NP_067649.2 | 10 | 852 | |||
| XM_005245404.3 | XP_005245461.1 | 10 | 909 | |||
| XM_011509838.2 | XP_011508140.1 | 9 | 838 | |||
| XM_011509839.2 | XP_011508141.1 | 10 | 911 | |||
| XM_011509840.2 | XP_011508142.1 | 9 | 694 | |||
| XM_011509841.2 | XP_011508143.1 | 7 | 589 | |||
| XM_011509842.2 | XP_011508144.1 | 7 | 2317 | |||
| XM_011509843.2 | XP_011508145.1 | 7 | 709 | |||
| XM_011509844.2 | XP_011508146.1 | 4 | 218 | |||
| XM_011509846.2 | XP_011508148.1 | 7 | 614 | |||
| XM_017001996.1 | XP_016857485.1 | 10 | 911 | |||
| XM_017001997.1 | XP_016857486.1 | 8 | 724 | |||
| GenBank mRNA | AB049405.1 | - | 10 | 850 | ||
| AF190501.1 | - | 8 | 548 | |||
| AK123055.1 | - | 8 | 795 | |||
| AY358119.1 | - | 10 | 893 | |||
| BC047905.1 | - | 10 | 835 | 5212508 | ||

LGR6
Genomic Map