siRNA Details
| Gene Symbol: AFAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134647.1 | NP_001128119.1 | 12 | 1705 | ||
| NM_198595.2 | NP_940997.1 | 12 | 1705 | |||
| XM_006713909.3 | XP_006713972.1 | 12 | 1588 | |||
| XM_011513544.2 | XP_011511846.1 | 12 | 1758 | |||
| XM_017008535.1 | XP_016864024.1 | 11 | 1434 | |||
| XM_017008536.1 | XP_016864025.1 | 11 | 1434 | |||
| GenBank mRNA | AB209676.1 | - | 9 | 1158 | ||
| AF188700.1 | - | 11 | 1432 | |||
| AK127962.1 | - | 3 | 427 | |||
| AK290807.1 | - | 12 | 1532 | |||
| AK297631.1 | - | 12 | 1498 | |||
| BC032777.1 | - | 11 | 1456 | 4914388 | ||

AFAP1
Genomic Map