siRNA Details
| Gene Symbol: GPBP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021639.4 | NP_067652.1 | 8 | 2029 | ||
| XM_017002019.1 | XP_016857508.1 | 8 | 2090 | |||
| GenBank mRNA | AF177334.1 | - | 6 | 847 | ||
| AK024972.1 | - | 6 | 836 | |||
| AK225197.1 | - | 6 | 836 | |||
| AL834194.1 | - | 6 | 1679 | |||
| BC015713.1 | - | 6 | 928 | 3909296 | ||
| BC022218.1 | - | 6 | 1112 | 3900818 | ||
| BC052257.1 | - | 5 | 677 | 6297561 | ||
| BC068578.1 | - | 8 | 2009 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_029868, BC031428, AK037224, AK133761, AK077148, and others.

GPBP1L1
Genomic Map