siRNA Details
| Gene Symbol: C12orf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021640.3 | NP_067653.3 | 5 | 750 | ||
| GenBank mRNA | AF111805.1 | - | 3 | 342 | ||
| AF289485.1 | - | 5 | 718 | |||
| BC013956.2 | - | 4 | 537 | |||
| BC028904.1 | - | 4 | 654 | 4454943 | ||
| BC051871.1 | - | 5 | 709 | 5170816 | ||
| BM551107.1 | - | 1 | 56 | |||

C12orf10
Genomic Map