siRNA Details
| Gene Symbol: COX7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001867.2 | NP_001858.1 | 2 | 237 | ||
| GenBank mRNA | AK026505.1 | - | 2 | 263 | ||
| BC001005.2 | - | 2 | 237 | 2821167 | ||
| BC007498.2 | - | 2 | 201 | 3030363 | ||
| BT007098.1 | - | 2 | 148 | |||
| X16560.1 | - | 2 | 166 | |||

COX7C
Genomic Map