siRNA Details
| Gene Symbol: EXOC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037126.1 | NP_001032203.1 | 4 | 604 | ||
| NM_021807.3 | NP_068579.3 | 4 | 604 | |||
| XM_005250523.4 | XP_005250580.1 | 4 | 588 | |||
| XM_017012495.1 | XP_016867984.1 | 4 | 588 | |||
| GenBank mRNA | AB051486.1 | - | 4 | 546 | ||
| AF380839.1 | - | 4 | 601 | |||
| AK027688.1 | - | 4 | 598 | |||
| AK295404.1 | - | 4 | 612 | |||
| AL831989.2 | - | 4 | 599 | |||
| BC026174.1 | - | 4 | 580 | |||
| BC067263.1 | - | 4 | 581 | 6183004 | ||

EXOC4
Genomic Map