siRNA Details
| Gene Symbol: CCDC90B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286116.1 | NP_001273045.1 | 2 | 363 | ||
| NM_001286117.1 | NP_001273046.1 | 2 | 910 | |||
| NM_001286118.1 | NP_001273047.1 | 2 | 590 | |||
| NM_001286119.1 | NP_001273048.1 | 3 | 675 | |||
| NM_021825.4 | NP_068597.2 | 2 | 590 | |||
| XM_005274154.2 | XP_005274211.1 | 2 | 849 | |||
| XM_005274156.3 | XP_005274213.1 | 3 | 681 | |||
| XM_005274159.3 | XP_005274216.1 | 3 | 256 | |||
| XM_017018103.1 | XP_016873592.1 | 2 | 292 | |||
| XM_017018104.1 | XP_016873593.1 | 3 | 681 | |||
| GenBank mRNA | AF182412.1 | - | 2 | 160 | ||
| AF182424.1 | - | 2 | 160 | |||
| AF271782.1 | - | 2 | 517 | |||
| AK055972.1 | - | 2 | 907 | |||
| AK225726.1 | - | 2 | 171 | |||
| AK304769.1 | - | 2 | 323 | |||
| AK315787.1 | - | 2 | 551 | |||
| AL136791.1 | - | 2 | 310 | |||
| AV756714.1 | - | 2 | 230 | |||
| BC014573.1 | - | 2 | 200 | 3680689 | ||
| BC020783.1 | - | 2 | 161 | 4773466 | ||
| BC067764.1 | - | 2 | 481 | |||
| BC107755.1 | - | 2 | 331 | 6737596 | ||
| BF131344.1 | - | 257 | ||||
| CN309108.1 | - | 2 | 590 | |||

CCDC90B
Genomic Map