siRNA Details
| Gene Symbol: RIC8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286134.1 | NP_001273063.1 | 2 | 1444 | ||
| NM_021932.5 | NP_068751.4 | 2 | 1444 | |||
| XM_005253052.3 | XP_005253109.1 | 2 | 630 | |||
| XM_017018107.1 | XP_016873596.1 | 2 | 714 | |||
| GenBank mRNA | AK026897.1 | - | 2 | 1443 | ||
| AK027461.1 | - | 2 | 133 | |||
| AK225420.1 | - | 2 | 133 | |||
| AK314568.1 | - | 2 | 154 | |||
| AL136935.1 | - | 2 | 425 | |||
| AL390088.1 | - | 2 | 165 | |||
| BC111499.1 | - | 2 | 261 | 6526180 | ||
| BC121807.1 | - | 2 | 341 | 40108083 | ||
| BC121808.1 | - | 2 | 341 | 40108080 | ||
| BX538115.1 | - | 1 | 218 | |||

RIC8A
Genomic Map