siRNA Details
| Gene Symbol: MIIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021933.3 | NP_068752.2 | 5 | 1004 | ||
| XM_005263487.3 | XP_005263544.1 | 5 | 833 | |||
| XM_011541895.1 | XP_011540197.1 | 5 | 835 | |||
| XM_011541896.1 | XP_011540198.1 | 5 | 728 | |||
| GenBank mRNA | AK022500.1 | - | 5 | 784 | ||
| AK024020.1 | - | 5 | 723 | |||
| AK095928.1 | - | 2 | 118 | |||
| BC008068.2 | - | 5 | 713 | 3050071 | ||
| FJ618905.1 | - | 5 | 1004 | |||

MIIP
Genomic Map