siRNA Details
| Gene Symbol: CELF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172673.1 | NP_001166144.1 | 11 | 1199 | ||
| NM_021938.3 | NP_068757.2 | 11 | 1316 | |||
| NR_033342.1 | 11 | 1113 | ||||
| XM_006722832.1 | XP_006722895.1 | 11 | 1343 | |||
| XM_006722833.1 | XP_006722896.1 | 11 | 1340 | |||
| XM_006722836.2 | XP_006722899.1 | 10 | 984 | |||
| XM_006722837.3 | XP_006722900.1 | 11 | 1255 | |||
| XM_006722838.3 | XP_006722901.1 | 10 | 1127 | |||
| XM_011528173.1 | XP_011526475.1 | 10 | 956 | |||
| XM_011528174.2 | XP_011526476.1 | 10 | 1125 | |||
| XM_017027111.1 | XP_016882600.1 | 9 | 1199 | |||
| GenBank mRNA | AB209067.1 | - | 11 | 1199 | ||
| AF248649.1 | - | 1 | 95 | |||
| AF329266.1 | - | 11 | 1279 | |||
| AK294109.1 | - | 11 | 1113 | |||
| AL832550.1 | - | 9 | 862 | |||
| BC028101.1 | - | 11 | 1316 | 5242909 | ||

CELF5
Genomic Map