siRNA Details
| Gene Symbol: CP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000096.3 | NP_000087.1 | 5 | 1163 | ||
| NR_046371.1 | 5 | 1163 | ||||
| XM_006713499.2 | XP_006713562.1 | 5 | 1168 | |||
| XM_006713500.3 | XP_006713563.1 | 5 | 1168 | |||
| XM_006713501.2 | XP_006713564.1 | 5 | 1168 | |||
| XM_011512435.1 | XP_011510737.1 | 5 | 1168 | |||
| XM_017005734.1 | XP_016861223.1 | 5 | 1168 | |||
| XM_017005735.1 | XP_016861224.1 | 5 | 1168 | |||
| GenBank mRNA | AK095290.1 | - | 4 | 452 | ||
| AK291219.1 | - | 5 | 945 | |||
| AK299272.1 | - | 5 | 949 | |||
| AK309279.1 | - | 1 | 135 | |||
| BC094709.1 | - | 5 | 947 | |||
| BC142714.1 | - | 5 | 919 | 40148840 | ||
| BC146663.1 | - | 5 | 919 | 40148048 | ||
| BC146801.1 | - | 5 | 943 | |||
| M13536.1 | - | 2 | 260 | |||
| M13699.1 | - | 5 | 910 | |||

CP
Genomic Map