siRNA Details
| Gene Symbol: THADA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083953.1 | NP_001077422.1 | 24 | 3947 | ||
| NM_022065.4 | NP_071348.3 | 24 | 3735 | |||
| NR_073394.1 | 24 | 3727 | ||||
| XM_006712061.3 | XP_006712124.1 | 25 | 3782 | |||
| XM_006712062.1 | XP_006712125.1 | 24 | 3732 | |||
| XM_006712063.1 | XP_006712126.1 | 24 | 3615 | |||
| XM_006712064.2 | XP_006712127.1 | 24 | 3735 | |||
| XM_006712065.1 | XP_006712128.1 | 24 | 3735 | |||
| XM_006712066.1 | XP_006712129.1 | 22 | 3372 | |||
| XM_006712067.1 | XP_006712130.1 | 22 | 3369 | |||
| XM_006712068.3 | XP_006712131.1 | 24 | 3735 | |||
| XM_006712069.3 | XP_006712132.1 | 9 | 1243 | |||
| XM_011533015.2 | XP_011531317.1 | 24 | 3735 | |||
| XM_017004674.1 | XP_016860163.1 | 24 | 3612 | |||
| XM_017004675.1 | XP_016860164.1 | 23 | 3487 | |||
| GenBank mRNA | AB051554.2 | - | 24 | 3947 | ||
| AK025445.1 | - | 11 | 1536 | |||
| AK126824.1 | - | 13 | 2133 | |||
| AK290023.1 | - | 24 | 3735 | |||
| AK307915.1 | - | 9 | 1060 | |||
| AL832141.1 | - | 24 | 3717 | |||
| AY149629.1 | - | 24 | 3727 | |||
| AY149633.1 | - | 24 | 3719 | |||
| BC105976.1 | - | 3 | 245 | |||
| BX648155.1 | - | 24 | 3731 | |||

THADA
Genomic Map