siRNA Details
| Gene Symbol: CPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001868.3 | NP_001859.1 | 2 | 238 | ||
| GenBank mRNA | AK131054.1 | - | 115 | |||
| AK291493.1 | - | 2 | 115 | |||
| AK293366.1 | - | 2 | 115 | |||
| BC005279.1 | - | 2 | 110 | 3949850 | ||
| BT007313.1 | - | 2 | 88 | |||
| X67318.1 | - | 2 | 95 | |||

CPA1
Genomic Map