siRNA Details
| Gene Symbol: HEATR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022070.4 | NP_071353.4 | 3 | 389 | ||
| XM_005257570.3 | XP_005257627.1 | 4 | 577 | |||
| XM_011525110.2 | XP_011523412.1 | 2 | 276 | |||
| XM_017024936.1 | XP_016880425.1 | 3 | 650 | |||
| XM_017024937.1 | XP_016880426.1 | 3 | 2326 | |||
| XM_017024938.1 | XP_016880427.1 | 3 | 379 | |||
| XM_017024939.1 | XP_016880428.1 | 3 | 650 | |||
| GenBank mRNA | AF349752.1 | - | 4 | 334 | ||
| AK127708.1 | - | 3 | 383 | |||
| AK314655.1 | - | 3 | 379 | |||
| BC119771.1 | - | 3 | 380 | 40108435 | ||
| BC119813.1 | - | 3 | 380 | 40108426 | ||
| BX640819.1 | - | 3 | 380 | |||
| BX640831.1 | - | 3 | 380 | |||
| CR627418.1 | - | 3 | 380 | |||
| DA375099.1 | - | 3 | 389 | |||

HEATR6
Genomic Map