siRNA Details
| Gene Symbol: ZBED8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303251.1 | NP_001290180.1 | 3 | 871 | ||
| NM_022090.4 | NP_071373.2 | 2 | 797 | |||
| GenBank mRNA | AK026754.1 | - | 1 | 603 | ||
| BC032603.1 | - | 3 | 818 | 5535434 | ||
| BC112128.1 | - | 1 | 339 | 8327596 | ||
| BC112130.1 | - | 1 | 339 | 8327598 | ||
| BM457021.1 | - | 2 | 751 | |||
| BQ959342.1 | - | 2 | 734 | |||

ZBED8
Genomic Map