siRNA Details
| Gene Symbol: CHP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022097.3 | NP_071380.1 | 6, 7 | 956 | ||
| GenBank mRNA | AF146019.1 | - | 6, 7 | 961 | ||
| AK290253.1 | - | 6, 7 | 542 | |||
| AK313062.1 | - | 6, 7 | 812 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHP2| Gene Symbol: CHP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022097.3 | NP_071380.1 | 6, 7 | 956 | ||
| GenBank mRNA | AF146019.1 | - | 6, 7 | 961 | ||
| AK290253.1 | - | 6, 7 | 542 | |||
| AK313062.1 | - | 6, 7 | 812 | |||