siRNA Details
| Gene Symbol: XPNPEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022098.3 | NP_071381.1 | 6 | 990 | ||
| GenBank mRNA | AK313770.1 | - | 6 | 951 | ||
| AL365514.1 | - | 6 | 973 | |||
| AL834310.1 | - | 7 | 1032 | |||
| BC001681.2 | - | 6 | 957 | 2823656 | ||
| BC004989.1 | - | 6 | 957 | 2823656 | ||
| BX648018.1 | - | 6 | 985 | |||
| CR456442.1 | - | 6 | 926 | |||
| CR457396.1 | - | 6 | 889 | |||

XPNPEP3
Genomic Map