siRNA Details
| Gene Symbol: CXorf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170569.1 | NP_001164040.1 | 5, 6 | 684 | ||
| NM_001170570.1 | NP_001164041.1 | 4, 5 | 560 | |||
| NM_022101.3 | NP_071384.1 | 5, 6 | 602 | |||
| GenBank mRNA | AK096297.1 | - | 5, 6 | 648 | ||
| AK298877.1 | - | 4, 5 | 538 | |||
| BC023506.2 | - | 5, 6 | 563 | |||
| BQ223940.1 | - | 5, 6 | 567 | |||

CXorf56
Genomic Map