siRNA Details
| Gene Symbol: CXorf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170569.1 | NP_001164040.1 | 3 | 419 | ||
| NM_022101.3 | NP_071384.1 | 3 | 337 | |||
| GenBank mRNA | AK096297.1 | - | 3 | 383 | ||
| BC023506.2 | - | 3 | 298 | |||
| BQ223940.1 | - | 3 | 302 | |||
| DA590933.1 | - | 3 | 421 | |||
| DA921725.1 | - | 3 | 312 | |||
| DB033119.1 | - | 3 | 352 | |||

CXorf56
Genomic Map