siRNA Details
| Gene Symbol: FAM217B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190826.1 | NP_001177755.1 | 5 | 843 | ||
| NM_022106.2 | NP_071389.1 | 4 | 777 | |||
| XM_011528985.2 | XP_011527287.1 | 4 | 767 | |||
| XM_011528986.2 | XP_011527288.1 | 5 | 854 | |||
| GenBank mRNA | AK125073.1 | - | 5 | 843 | ||
| AL137442.1 | - | 1 | 227 | |||
| BC020771.1 | - | 4 | 696 | |||
| BC054002.1 | - | 4 | 665 | 6066745 | ||

FAM217B
Genomic Map