siRNA Details
| Gene Symbol: DMRTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032110.2 | NP_115486.1 | 3 | 1846 | ||
| XM_011541937.2 | XP_011540239.1 | 2 | 2635 | |||
| GenBank mRNA | AJ301580.1 | - | 2 | 1340 | ||
| BC136282.1 | - | 2 | 1590 | 9020269 | ||
| BC143800.1 | - | 2 | 1596 | 9052314 | ||

DMRTA2
Genomic Map