siRNA Details
| Gene Symbol: DMRTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032110.2 | NP_115486.1 | 3 | 1851 | ||
| XM_011541937.2 | XP_011540239.1 | 2 | 2640 | |||
| GenBank mRNA | BC136282.1 | - | 2 | 1595 | 9020269 | |
| BC143800.1 | - | 2 | 1601 | 9052314 | ||

DMRTA2
Genomic Map