siRNA Details
| Gene Symbol: CLSPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190481.1 | NP_001177410.1 | 12 | 2257 | ||
| NM_022111.3 | NP_071394.2 | 13 | 2449 | |||
| XM_011541938.2 | XP_011540240.1 | 13 | 2472 | |||
| XM_011541939.2 | XP_011540241.1 | 13 | 2466 | |||
| XM_011541940.2 | XP_011540242.1 | 13 | 2466 | |||
| XM_011541941.2 | XP_011540243.1 | 12 | 2304 | |||
| GenBank mRNA | AF297866.1 | - | 13 | 2449 | ||
| BC113116.1 | - | 13 | 2442 | 7961377 | ||
| BC115025.1 | - | 12 | 2250 | 7961363 | ||
| BC115026.1 | - | 13 | 2442 | 7961366 | ||
| BC137279.1 | - | 13 | 2428 | 9021276 | ||
| BC140789.1 | - | 13 | 2428 | 9021657 | ||

CLSPN
Genomic Map