siRNA Details
| Gene Symbol: NEUROG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024019.3 | NP_076924.1 | 2 | 649 | ||
| GenBank mRNA | BC036847.1 | - | 2 | 649 | 5247719 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009718, Y07621, BC055743, AK143190.

NEUROG2
Genomic Map