siRNA Details
| Gene Symbol: SNX16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022133.3 | NP_071416.2 | 3 | 828 | ||
| NM_152836.2 | NP_690049.1 | 2 | 582 | |||
| NM_152837.2 | NP_690050.1 | 2 | 582 | |||
| XM_005251282.4 | XP_005251339.1 | 3 | 535 | |||
| XM_005251283.2 | XP_005251340.1 | 4 | 672 | |||
| GenBank mRNA | AF305779.1 | - | 2 | 433 | ||
| AK290903.1 | - | 2 | 498 | |||
| AL833763.1 | - | 2 | 489 | |||
| BC033630.1 | - | 2 | 490 | 5224216 | ||
| BG739930.1 | - | 2 | 496 | |||
| CB962009.1 | - | 2 | 560 | |||

SNX16
Genomic Map