siRNA Details
| Gene Symbol: GAL3ST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022134.2 | NP_071417.2 | 3, 4 | 509 | ||
| GenBank mRNA | AB040610.1 | - | 3, 4 | 510 | ||
| BC117293.1 | - | 3, 4 | 427 | 40125844 | ||
| BC117295.1 | - | 3, 4 | 427 | 40125846 | ||

GAL3ST2
Genomic Map