siRNA Details
| Gene Symbol: LRRC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022143.4 | NP_071426.1 | 2 | 1942 | ||
| XM_011516461.2 | XP_011514763.1 | 3 | 2248 | |||
| XM_017012505.1 | XP_016867994.1 | 3 | 2138 | |||
| GenBank mRNA | AF196976.4 | - | 2 | 1945 | ||
| AJ297858.1 | - | 2 | 1933 | |||
| AK172751.1 | - | 2 | 1946 | |||
| AK314047.1 | - | 2 | 2018 | |||
| AY358307.1 | - | 1 | 1861 | |||
| BC111561.1 | - | 2 | 1936 | 40068142 | ||
| BC111745.1 | - | 2 | 1936 | 40068146 | ||

LRRC4
Genomic Map