siRNA Details
| Gene Symbol: CENPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267038.1 | NP_001253967.1 | 8 | 586 | ||
| NM_022145.4 | NP_071428.2 | 8 | 620 | |||
| XM_011543534.2 | XP_011541836.1 | 8 | 564 | |||
| XM_011543535.2 | XP_011541837.1 | 8 | 682 | |||
| XM_011543536.2 | XP_011541838.1 | 9 | 842 | |||
| XM_011543537.2 | XP_011541839.1 | 8 | 598 | |||
| XM_011543540.2 | XP_011541842.1 | 8 | 564 | |||
| XM_011543541.2 | XP_011541843.1 | 5 | 366 | |||
| XM_011543542.2 | XP_011541844.1 | 5 | 486 | |||
| XM_017009690.1 | XP_016865179.1 | 8 | 825 | |||
| XM_017009691.1 | XP_016865180.1 | 8 | 576 | |||
| XM_017009692.1 | XP_016865181.1 | 8 | 794 | |||
| XM_017009693.1 | XP_016865182.1 | 8 | 564 | |||
| XM_017009694.1 | XP_016865183.1 | 9 | 793 | |||
| XM_017009695.1 | XP_016865184.1 | 7 | 462 | |||
| XM_017009696.1 | XP_016865185.1 | 8 | 564 | |||
| XM_017009697.1 | XP_016865186.1 | 7 | 496 | |||
| XM_017009699.1 | XP_016865188.1 | 8 | 598 | |||
| XM_017009700.1 | XP_016865189.1 | 7 | 2930 | |||
| XM_017009701.1 | XP_016865190.1 | 6 | 561 | |||
| GenBank mRNA | AF315941.1 | - | 8 | 576 | ||
| AY009151.1 | - | 6 | 450 | |||
| BC005400.1 | - | 8 | 576 | 3839409 | ||
| BC008504.1 | - | 8 | 620 | 3511065 | ||

CENPK
Genomic Map