siRNA Details
| Gene Symbol: MAGEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022149.4 | NP_071432.2 | 1 | 667 | ||
| GenBank mRNA | AF295378.1 | - | 1 | 616 | ||
| AF320910.1 | - | 1 | 535 | |||
| BC010056.2 | - | 1 | 621 | 3943384 | ||
| BQ058163.1 | - | 1 | 273 | |||
| BQ070043.1 | - | 1 | 668 | |||

MAGEF1
Genomic Map