siRNA Details
| Gene Symbol: FN3K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022158.3 | NP_071441.1 | 2 | 320 | ||
| GenBank mRNA | AJ404615.1 | - | 2 | 280 | ||
| AK054838.1 | - | 2 | 1270 | |||
| AK294871.1 | - | 2 | 281 | |||
| BC042680.1 | - | 2 | 264 | 5195472 | ||

FN3K
Genomic Map