siRNA Details
| Gene Symbol: RBSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302378.1 | NP_001289307.1 | 5 | 845 | ||
| NM_022340.3 | NP_071735.2 | 6 | 1001 | |||
| XM_005265384.4 | XP_005265441.1 | 6 | 964 | |||
| XM_005265385.4 | XP_005265442.1 | 5 | 819 | |||
| XM_011534001.2 | XP_011532303.1 | 2 | 341 | |||
| XM_017007023.1 | XP_016862512.1 | 5 | 837 | |||
| XM_017007024.1 | XP_016862513.1 | 7 | 1147 | |||
| XM_017007025.1 | XP_016862514.1 | 6 | 1006 | |||
| XM_017007027.1 | XP_016862516.1 | 6 | 992 | |||
| GenBank mRNA | AB209673.1 | - | 6 | 961 | ||
| AK291288.1 | - | 5 | 836 | |||
| AK301735.1 | - | 5 | 837 | |||
| AL833545.1 | - | 6 | 995 | |||
| AY009133.1 | - | 3 | 378 | |||
| BC106940.2 | - | 3 | 378 | 40034008 | ||

RBSN
Genomic Map