siRNA Details
| Gene Symbol: C17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022344.3 | NP_071739.2 | 10 | 1146 | ||
| XM_005258022.3 | XP_005258079.1 | 11 | 1164 | |||
| XM_017024940.1 | XP_016880429.1 | 10 | 1015 | |||
| XM_017024941.1 | XP_016880430.1 | 10 | 1013 | |||
| GenBank mRNA | AB062437.1 | - | 4 | 368 | ||
| BC026017.2 | - | 10 | 1092 | 3352761 | ||
| BC035715.1 | - | 10 | 1100 | 5588164 | ||
| BC047325.1 | - | 10 | 1086 | 5493477 | ||
| BC054035.1 | - | 10 | 1080 | 6143106 | ||

C17orf75
Genomic Map