siRNA Details
| Gene Symbol: IRF2BPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024496.3 | NP_078772.1 | 1 | 3081 | ||
| GenBank mRNA | AB058768.1 | - | 2 | 2576 | ||
| AF063597.1 | - | 1 | 211 | |||
| AL832320.1 | - | 1 | 1183 | |||
| BC108292.1 | - | 1 | 3078 | |||
| BC152431.1 | - | 2 | 2576 | |||

IRF2BPL
Genomic Map