siRNA Details
| Gene Symbol: DNAJC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022365.3 | NP_071760.2 | 7 | 1075 | ||
| XM_011519614.2 | XP_011517916.1 | 7 | 1122 | |||
| XM_017016536.1 | XP_016872025.1 | 7 | 1122 | |||
| GenBank mRNA | AK026062.1 | - | 4 | 348 | ||
| AK027263.1 | - | 7 | 895 | |||
| AK308369.1 | - | 7 | 1081 | |||
| AY225122.1 | - | 7 | 788 | |||
| BC030955.1 | - | 3 | 258 | |||
| BC110894.1 | - | 7 | 1075 | 5164559 | ||

DNAJC1
Genomic Map