siRNA Details
| Gene Symbol: DNAJC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022365.3 | NP_071760.2 | 9 | 1370 | ||
| XM_011519614.2 | XP_011517916.1 | 9 | 1417 | |||
| GenBank mRNA | AF427142.1 | - | 2 | 246 | ||
| AK026062.1 | - | 6 | 643 | |||
| AK027263.1 | - | 9 | 1190 | |||
| AY225122.1 | - | 9 | 1083 | |||
| BC030955.1 | - | 5 | 553 | |||
| BC110894.1 | - | 9 | 1370 | 5164559 | ||
| BC139752.1 | - | 1 | 90 | |||

DNAJC1
Genomic Map